Assay Details
| Gene Symbol: nac-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027502.4 | NP_001022673.1 | 1 - 2 | 165 | 58 | |
| GenBank mRNA | AY090485.1 | - | 1 - 2 | 162 | 58 | |
More Information
Set Membership: |
|

nac-3
Genomic Map