Assay Details
| Gene Symbol: mua-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027503.4 | NP_001022674.1 | 7 - 8 | 1246 | 120 | |
| GenBank mRNA | AF139060.1 | - | 7 - 8 | 1246 | 120 | |
More Information
Set Membership: |
|

mua-3
Genomic Map