Assay Details
| Gene Symbol: unc-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066878.4 | NP_499279.2 | 2 - 3 | 351 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

unc-50| Gene Symbol: unc-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066878.4 | NP_499279.2 | 2 - 3 | 351 | 77 | |
Set Membership: |
|