Assay Details
| Gene Symbol: M03C11.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066900.7 | NP_499301.2 | 6 - 7 | 1485 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

M03C11.8| Gene Symbol: M03C11.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066900.7 | NP_499301.2 | 6 - 7 | 1485 | 76 | |
Set Membership: |
|