Assay Details
| Gene Symbol: atx-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379897.1 | NP_001367241.1 | 9 - 10 | 2609 | 87 | |
| NM_001379898.1 | NP_001366696.1 | 9 - 10 | 2370 | 87 | ||
| NM_001379899.1 | NP_001367186.1 | 8 - 9 | 2285 | 87 | ||
| NM_001379900.2 | NP_001367113.1 | 9 - 10 | 2258 | 87 | ||
More Information
Set Membership: |
|

atx-2
Genomic Map