Assay Details
| Gene Symbol: atx-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379897.1 | NP_001367241.1 | 11 - 12 | 3111 | 63 | |
| NM_001379898.1 | NP_001366696.1 | 11 - 12 | 2872 | 63 | ||
| NM_001379900.2 | NP_001367113.1 | 11 - 12 | 2760 | 63 | ||
More Information
Set Membership: |
|

atx-2
Genomic Map