Assay Details
| Gene Symbol: atx-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379897.1 | NP_001367241.1 | 5 - 6 | 1300 | 69 | |
| NM_001379898.1 | NP_001366696.1 | 5 - 6 | 1061 | 69 | ||
| NM_001379899.1 | NP_001367186.1 | 4 - 5 | 976 | 69 | ||
| NM_001379900.2 | NP_001367113.1 | 5 - 6 | 949 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379900.2 transcripts.
Set Membership: |
|

atx-2
Genomic Map