Assay Details
| Gene Symbol: unc-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027611.3 | NP_001022782.1 | 7 - 8 | 1019 | 108 | |
| NM_001379903.1 | NP_001368253.1 | 3 - 4 | 392 | 108 | ||
| GenBank mRNA | AF151640.1 | - | 7 - 8 | 1188 | 108 | |
More Information
Set Membership: |
|

unc-49
Genomic Map