Assay Details
| Gene Symbol: unc-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027611.3 | NP_001022782.1 | 20 - 21 | 2636 | 124 | |
| NM_001027612.4 | NP_001022783.1 | 13 - 14 | 1819 | 124 | ||
| NM_001027613.3 | NP_001022784.1 | 12 - 13 | 1783 | 124 | ||
| NM_001027614.4 | NP_001022785.1 | 14 - 15 | 1795 | 124 | ||
| NM_001027615.4 | NP_001022786.1 | 6 - 7 | 918 | 124 | ||
| NM_001306734.3 | NP_001293663.1 | 2 - 3 | 208 | 124 | ||
| NM_001379901.2 | NP_001368252.1 | 12 - 13 | 2366 | 124 | ||
| NM_001379904.1 | NP_001367142.1 | 12 - 13 | 2447 | 124 | ||
| GenBank mRNA | AF151641.1 | - | 13 - 14 | 1745 | 124 | |
| AF151642.1 | - | 12 - 13 | 1709 | 124 | ||
| AF151643.1 | - | 14 - 15 | 1802 | 124 | ||
| AF151644.1 | - | 6 - 7 | 825 | 124 | ||
| AF151645.1 | - | 1 - 2 | 324 | 124 | ||
More Information
Set Membership: |
|

unc-49
Genomic Map