Assay Details
| Gene Symbol: unc-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027614.4 | NP_001022785.1 | 8 - 9 | 1247 | 93 | |
| GenBank mRNA | AF151643.1 | - | 8 - 9 | 1254 | 93 | |
More Information
Set Membership: |
|

unc-49
Genomic Map