Assay Details
| Gene Symbol: tat-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066962.5 | NP_499363.3 | 4 - 5 | 1318 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tat-3| Gene Symbol: tat-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066962.5 | NP_499363.3 | 4 - 5 | 1318 | 108 | |
Set Membership: |
|