Assay Details
| Gene Symbol: ptr-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268156.3 | NP_001255085.1 | 7 - 8 | 917 | 101 | |
| NM_001268157.3 | NP_001255086.1 | 7 - 8 | 889 | 101 | ||
| NM_001268158.3 | NP_001255087.1 | 7 - 8 | 889 | 101 | ||
More Information
Set Membership: |
|

ptr-19
Genomic Map