Assay Details
| Gene Symbol: sca-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066984.8 | NP_499385.3 | 4 - 5 | 366 | 80 | |
| NM_066985.6 | NP_499386.1 | 4 - 5 | 412 | 80 | ||
| GenBank mRNA | AJ012296.1 | - | 3 - 4 | 386 | 80 | |
More Information
Set Membership: |
|

sca-1
Genomic Map