Assay Details
| Gene Symbol: vps-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066993.3 | NP_499394.2 | 1 - 2 | 271 | 136 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

vps-50| Gene Symbol: vps-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066993.3 | NP_499394.2 | 1 - 2 | 271 | 136 | |
Set Membership: |
|