Assay Details
| Gene Symbol: smc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268189.3 | NP_001255118.1 | 3 - 4 | 397 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smc-3| Gene Symbol: smc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268189.3 | NP_001255118.1 | 3 - 4 | 397 | 121 | |
Set Membership: |
|