Assay Details
| Gene Symbol: rab-35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067053.6 | NP_499454.1 | 3 - 4 | 382 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rab-35| Gene Symbol: rab-35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067053.6 | NP_499454.1 | 3 - 4 | 382 | 64 | |
Set Membership: |
|