Assay Details
| Gene Symbol: mcm-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067055.4 | NP_499456.1 | 3 - 4 | 589 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mcm-10| Gene Symbol: mcm-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067055.4 | NP_499456.1 | 3 - 4 | 589 | 132 | |
Set Membership: |
|