Assay Details
| Gene Symbol: dpy-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067063.5 | NP_499464.1 | 7 - 8 | 1630 | 61 | |
| GenBank mRNA | AJ270999.1 | - | 6 - 7 | 1512 | 61 | |
More Information
Set Membership: |
|

dpy-18
Genomic Map