Assay Details
| Gene Symbol: xpb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067086.5 | NP_499487.1 | 6 - 7 | 2323 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

xpb-1| Gene Symbol: xpb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067086.5 | NP_499487.1 | 6 - 7 | 2323 | 71 | |
Set Membership: |
|