Assay Details
| Gene Symbol: gbf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379938.1 | NP_001366879.1 | 15 - 16 | 4709 | 62 | |
| NM_001379939.1 | NP_001368124.1 | 1 - 2 | 219 | 62 | ||
More Information
Set Membership: |
|

gbf-1
Genomic Map