Assay Details
| Gene Symbol: ntl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083183.3 | NP_001076652.1 | 7 - 8 | 1941 | 58 | |
| NM_001083184.5 | NP_001076653.1 | 1 - 2 | 290 | 58 | ||
More Information
Set Membership: |
|

ntl-3
Genomic Map