Assay Details
| Gene Symbol: ccf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067152.7 | NP_499553.1 | 2 - 3 | 432 | 75 | |
| GenBank mRNA | U21854.1 | - | 2 - 3 | 442 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ccf-1| Gene Symbol: ccf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067152.7 | NP_499553.1 | 2 - 3 | 432 | 75 | |
| GenBank mRNA | U21854.1 | - | 2 - 3 | 442 | 75 | |
Set Membership: |
|