Assay Details
| Gene Symbol: top-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067157.6 | NP_499558.1 | 5 - 6 | 1113 | 98 | |
| GenBank mRNA | AF057032.1 | - | 5 - 6 | 1113 | 98 | |
More Information
Set Membership: |
|

top-3
Genomic Map