Assay Details
| Gene Symbol: ung-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268219.3 | NP_001255148.1 | 1 - 2 | 602 | 98 | |
| NM_067159.6 | NP_499560.1 | 2 - 3 | 735 | 98 | ||
More Information
Set Membership: |
|

ung-1
Genomic Map