Assay Details
| Gene Symbol: php-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067172.6 | NP_499573.1 | 2 - 3 | 647 | 62 | |
| GenBank mRNA | AF172092.1 | - | 1 - 2 | 405 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

php-3| Gene Symbol: php-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067172.6 | NP_499573.1 | 2 - 3 | 647 | 62 | |
| GenBank mRNA | AF172092.1 | - | 1 - 2 | 405 | 62 | |
Set Membership: |
|