Assay Details
| Gene Symbol: php-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067172.6 | NP_499573.1 | 4 - 5 | 1033 | 81 | |
| GenBank mRNA | AF172092.1 | - | 3 - 4 | 791 | 81 | |
More Information
Set Membership: |
|

php-3
Genomic Map