Assay Details
| Gene Symbol: cest-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067175.6 | NP_499576.4 | 7 - 8 | 3379 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cest-12| Gene Symbol: cest-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067175.6 | NP_499576.4 | 7 - 8 | 3379 | 92 | |
Set Membership: |
|