Assay Details
| Gene Symbol: grl-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067191.8 | NP_499592.1 | 1 - 2 | 604 | 91 | |
| Gene Symbol: Y75B8A.18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067189.7 | NP_499590.2 | 1 - 2 | 266 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_067189.7 transcripts.
Set Membership: |
|

grl-15
Genomic Map