Assay Details
| Gene Symbol: tim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067193.7 | NP_499594.2 | 7 - 8 | 4045 | 80 | |
| GenBank mRNA | AF183401.1 | - | 6 - 7 | 3903 | 80 | |
More Information
Set Membership: |
|

tim-1
Genomic Map