Assay Details
| Gene Symbol: zip-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383010.1 | 5 - 6 | 531 | 89 | ||
| NM_067201.6 | NP_499602.3 | 4 - 5 | 581 | 89 | ||
| GenBank mRNA | AF160492.1 | - | 4 - 5 | 438 | 89 | |
More Information
Set Membership: |
|

zip-1
Genomic Map