Assay Details
| Gene Symbol: pph-4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027727.5 | NP_001022898.1 | 2 - 3 | 381 | 61 | |
| NM_001027728.5 | NP_001022899.1 | 1 - 2 | 62 | 61 | ||
More Information
Set Membership: |
|

pph-4.2
Genomic Map