Assay Details
| Gene Symbol: tat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027723.3 | NP_001022894.1 | 2 - 3 | 344 | 159 | |
| NM_001268236.3 | NP_001255165.1 | 2 - 3 | 196 | 159 | ||
| NM_001379952.1 | NP_001367088.1 | 1 - 2 | 41 | 159 | ||
More Information
Set Membership: |
|

tat-1
Genomic Map