Assay Details
| Gene Symbol: tat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027723.3 | NP_001022894.1 | 5 - 6 | 696 | 82 | |
| NM_001268236.3 | NP_001255165.1 | 5 - 6 | 548 | 82 | ||
| NM_001379952.1 | NP_001367088.1 | 4 - 5 | 393 | 82 | ||
More Information
Set Membership: |
|

tat-1
Genomic Map