Assay Details
| Gene Symbol: epc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383013.1 | 2 - 3 | 369 | 78 | ||
| GenBank mRNA | AY551964.1 | - | 1 - 2 | 286 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

epc-1| Gene Symbol: epc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383013.1 | 2 - 3 | 369 | 78 | ||
| GenBank mRNA | AY551964.1 | - | 1 - 2 | 286 | 78 | |
Set Membership: |
|