Assay Details
| Gene Symbol: epc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383013.1 | 4 - 5 | 726 | 85 | ||
| GenBank mRNA | AY551964.1 | - | 3 - 4 | 643 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

epc-1| Gene Symbol: epc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383013.1 | 4 - 5 | 726 | 85 | ||
| GenBank mRNA | AY551964.1 | - | 3 - 4 | 643 | 85 | |
Set Membership: |
|