Assay Details
| Gene Symbol: lgc-37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067261.7 | NP_499662.2 | 9 - 10 | 1315 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lgc-37| Gene Symbol: lgc-37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067261.7 | NP_499662.2 | 9 - 10 | 1315 | 83 | |
Set Membership: |
|