Assay Details
| Gene Symbol: snf-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067301.4 | NP_499702.2 | 6 - 7 | 1941 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

snf-7| Gene Symbol: snf-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067301.4 | NP_499702.2 | 6 - 7 | 1941 | 71 | |
Set Membership: |
|