Assay Details
| Gene Symbol: ife-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268267.4 | NP_001255196.1 | 1 - 2 | 395 | 140 | |
| Gene Symbol: dro-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379982.1 | NP_001366888.1 | 1 - 2 | 421 | 140 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001268267.4 transcripts.
Set Membership: |
|

ife-1
Genomic Map