Assay Details
| Gene Symbol: trf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067372.5 | NP_499773.2 | 10 - 11 | 1182 | 103 | |
| GenBank mRNA | AF348168.1 | - | 10 - 11 | 1107 | 103 | |
More Information
Set Membership: |
|

trf-1
Genomic Map