Assay Details
| Gene Symbol: trf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067372.5 | NP_499773.2 | 5 - 6 | 743 | 72 | |
| GenBank mRNA | AF348168.1 | - | 5 - 6 | 668 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

trf-1| Gene Symbol: trf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067372.5 | NP_499773.2 | 5 - 6 | 743 | 72 | |
| GenBank mRNA | AF348168.1 | - | 5 - 6 | 668 | 72 | |
Set Membership: |
|