Assay Details
| Gene Symbol: cdk-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067382.8 | NP_499783.1 | 3 - 4 | 679 | 111 | |
| GenBank mRNA | AF129111.1 | - | 3 - 4 | 584 | 111 | |
More Information
Set Membership: |
|

cdk-5
Genomic Map