Assay Details
| Gene Symbol: acs-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067398.6 | NP_499799.1 | 4 - 5 | 504 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acs-5| Gene Symbol: acs-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067398.6 | NP_499799.1 | 4 - 5 | 504 | 67 | |
Set Membership: |
|