Assay Details
Gene Symbol: Y76A2B.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067400.11 | NP_499801.1 | 3 - 4 | 209 | 79 |
Gene Symbol: Y76A2B.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067399.3 | NP_499800.1 | 1 - 2 | 58 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_067399.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
