Assay Details
| Gene Symbol: set-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067418.6 | NP_499819.3 | 12 - 13 | 6117 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

set-16| Gene Symbol: set-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067418.6 | NP_499819.3 | 12 - 13 | 6117 | 85 | |
Set Membership: |
|