Assay Details
| Gene Symbol: lit-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027634.3 | NP_001022805.1 | 1 - 2 | 145 | 61 | |
| GenBank mRNA | AF145376.1 | - | 1 - 2 | 145 | 61 | |
More Information
Set Membership: |
|

lit-1
Genomic Map