Assay Details
| Gene Symbol: lit-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027634.3 | NP_001022805.1 | 5 - 6 | 643 | 75 | |
| NM_001027635.5 | NP_001022806.1 | 3 - 4 | 308 | 75 | ||
| NM_001027637.5 | NP_001022808.1 | 6 - 7 | 675 | 75 | ||
| NM_001379989.1 | NP_001367102.1 | 1 - 2 | 103 | 75 | ||
| GenBank mRNA | AF143243.1 | - | 4 - 5 | 486 | 75 | |
| AF143244.1 | - | 2 - 3 | 174 | 75 | ||
| AF145376.1 | - | 5 - 6 | 643 | 75 | ||
More Information
Set Membership: |
|

lit-1
Genomic Map