Assay Details
| Gene Symbol: lit-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027634.3 | NP_001022805.1 | 9 - 10 | 1691 | 61 | |
| NM_001027635.5 | NP_001022806.1 | 7 - 8 | 1356 | 61 | ||
| NM_001027636.3 | NP_001022807.1 | 5 - 6 | 1567 | 61 | ||
| NM_001027637.5 | NP_001022808.1 | 10 - 11 | 1723 | 61 | ||
| NM_001379989.1 | NP_001367102.1 | 5 - 6 | 1151 | 61 | ||
| GenBank mRNA | AF143243.1 | - | 8 - 9 | 1534 | 61 | |
| AF143244.1 | - | 6 - 7 | 1222 | 61 | ||
| AF145376.1 | - | 9 - 10 | 1691 | 61 | ||
More Information
Set Membership: |
|

lit-1
Genomic Map