Assay Details
| Gene Symbol: pdr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306674.3 | NP_001293603.1 | 2 - 3 | 458 | 76 | |
| NM_001383028.1 | 3 - 4 | 656 | 76 | |||
| GenBank mRNA | AX254400.1 | - | 3 - 4 | 644 | 76 | |
More Information
Set Membership: |
|

pdr-1
Genomic Map