Assay Details
| Gene Symbol: mdt-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067446.7 | NP_499847.1 | 4 - 5 | 763 | 65 | |
| Gene Symbol: pdr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306674.3 | NP_001293603.1 | 3 - 4 | 550 | 65 | |
| NM_001383028.1 | 4 - 5 | 748 | 65 | |||
| GenBank mRNA | AX254400.1 | - | 4 - 5 | 736 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_067446.7 transcripts.
Set Membership: |
|

mdt-29
Genomic Map