Assay Details
| Gene Symbol: cox-6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067466.6 | NP_499867.1 | 2 - 3 | 193 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cox-6C| Gene Symbol: cox-6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067466.6 | NP_499867.1 | 2 - 3 | 193 | 75 | |
Set Membership: |
|